Learn More
Ace Glass, Inc. Tubing connection ferrule only 3.3mm 1/8"
SDP
Catalog No. NC0260533
Encompass
3.3m chemically inert tubing connector ferrule, with male -28 thread for connecting 1/8 O.D. PTFE or Tefzel tubing to chromatography adapters. Ferrule is made of pure, virgin Tefzel (ETFE); no coloring agents are added. When used with OPTI-LOK nut, it offers a zero-dead volume, leak-free seal.
Catalog No. NC0260533 Supplier Ace Glass, Inc. Supplier No. 585426
3.3m chemically inert tubing connector ferrule, with male -28 thread for connecting 1/8 O.D. PTFE or Tefzel tubing to chromatography adapters. Ferrule is made of pure, virgin Tefzel (ETFE); no coloring agents are added. When used with OPTI-LOK nut, it offers a zero-dead volume, leak-free seal.