Learn More
Ace Glass, Inc. Tubing connection nut only 3.3m 1/8"
SDP
Catalog No. NC0258249
Encompass
3.3m chemically inert tubing connector nut, with male -28 thread for connecting 1/8 O.D. PTFE or Tefzel tubing to chromatography adapters. Male connector nut is made of white acetal and nylon. When used with OPTI-LOK ferrule, it offers a zero-dead volume, leak-free seal.
Catalog No. NC0258249 Supplier Ace Glass, Inc. Supplier No. 585409
3.3m chemically inert tubing connector nut, with male -28 thread for connecting 1/8 O.D. PTFE or Tefzel tubing to chromatography adapters. Male connector nut is made of white acetal and nylon. When used with OPTI-LOK ferrule, it offers a zero-dead volume, leak-free seal.