Learn More
Bel-Art™ Complete Fittings Assortment
SCI_ED
Catalog No. S50710 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Includes:
Assortment of 72 plastic fittings
Catalog No. Includes
S50710 Assortment of 72 plastic fittings
1 options
Catalog No. S50710 Supplier Bel-Art™ Supplier No. F196250000/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Reduces time spent searching for the right tubing connector or stopper
This generous assortment of 72 plastic fittings has everything needed for routine laboratory set-ups.
  • Clear, hinge-top, polystyrene storage box makes it easy to find what you need
  • Includes commonly used sizes of connector and stopper styles
  • FDA grade, autoclavable polypropylene connectors: 15 Straight Tubing Connectors, 11 Stepped Tubing Connectors, 6 T-Shaped Tubing Connectors, 10 Wye (Y) Tubing Connectors
  • Polyethylene connectors: 3 pair Quick Disconnects
  • Polyethylene stoppers: 24 Cup-Type
Specifications
Autoclavable Autoclavable
Material Polypropylene, Polyethylene
Includes Assortment of 72 plastic fittings
Color Natural
Product Type Fittings Assortment