Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, 1/16th, SS, Hex Heads, Medium Length 10-32 Threads. Ferrules sold separately. 1 EA.
Catalog No. 502271583
Encompass_Preferred
These "Male Nuts" used to connect Stainless Steel tubing in the high pressures zones of HPLC systems. This 316L Stainless Steel Fitting has a standard diameter Hexagonal head for wrenching the nut. The medium thread length makes this Nut ideal for use on injectors such as the Rheodyne™ and others, where space is limited. Use with Single Cone Stainless Steel Ferrules which are designed to work with this fitting, providing a permanent attachment to SS tubing for a high pressure connection.
This item is not returnable. View return policy
Catalog No. 50-227-1583 Supplier Microsolv Technology Corporation Supplier No. 4959005
These "Male Nuts" used to connect Stainless Steel tubing in the high pressures zones of HPLC systems. This 316L Stainless Steel Fitting has a standard diameter Hexagonal head for wrenching the nut. The medium thread length makes this Nut ideal for use on injectors such as the Rheodyne™ and others, where space is limited. Use with Single Cone Stainless Steel Ferrules which are designed to work with this fitting, providing a permanent attachment to SS tubing for a high pressure connection.