Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, One Piece, 1/16", PEEK, Natural, Standard Size Head but a Long Length with 10-32 Screw Threads. Finger Tighten, 10/PK.
Catalog No. 502271480
Encompass_Preferred
These all PEEK "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. Fitting has a 12mm diameter knurled head, 25mm total length (top to bottom) with an 16mm thread length. The long length is useful when your column or union or detector has a deep threaded fitting and the extra length is needed to seat the tubing properly. The beveled inlet hole helps to make threading the tubing through the fitting easier.
This item is not returnable. View return policy
Catalog No. 50-227-1480 Supplier Microsolv Technology Corporation Supplier No. 4732004
These all PEEK "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. Fitting has a 12mm diameter knurled head, 25mm total length (top to bottom) with an 16mm thread length. The long length is useful when your column or union or detector has a deep threaded fitting and the extra length is needed to seat the tubing properly. The beveled inlet hole helps to make threading the tubing through the fitting easier.