Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, One Piece, 1/16", PEEK, Natural, Small Hex Head, Long Length and 10-32 Screw Threads. Wrench Tighten, 10/PK.
Catalog No. 502271495
Encompass_Preferred
This "Male Nut" is used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. This Fitting has a 7mm diameter hexagonal head for use with a wrench to tighten or loosen the nut. The smaller, 7mm diameter head allows this fitting to be used inside instruments when space is limited. The 20mm total length (top to bottom) with a 15mm thread length allows for a an excellent, swaged connection that is similar in use to a Stainless Steel nut.
This item is not returnable. View return policy
Catalog No. 50-227-1495 Supplier Microsolv Technology Corporation Supplier No. 4732021
This "Male Nut" is used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. This Fitting has a 7mm diameter hexagonal head for use with a wrench to tighten or loosen the nut. The smaller, 7mm diameter head allows this fitting to be used inside instruments when space is limited. The 20mm total length (top to bottom) with a 15mm thread length allows for a an excellent, swaged connection that is similar in use to a Stainless Steel nut.