Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, Two Pieces, 1/16", PEEK, Natural, Large Head, Standard Length and 10-32 Screw Threads. Use with Double Cone Ferrules. Finger Tighten, 1 EA.
Supplier:  Microsolv Technology Corporation 4732030
Encompass_Preferred
These Two Piece, "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. This fitting has a 12mm diameter knurled head, 18mm total length (top to bottom) and requires a Double Cone, Ferrule, sold separately. 2 Piece Male Nuts are often chosen because they can be used at slightly higher pressures compared to 1 Piece Nuts. The required, Double Coned Ferrules will swage tubing at two places and have a greater grip than one piece fittings. The larger Head makes it easier to produce the torque needed to achieve a good connection. As Ferrules wear down, they need to be replaced. This Nut can be reused with new Ferrules.
Catalog No. 50-227-1499