Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, Two Pieces, 1/16", PEEK, Natural, Large Head, Standard Length and 10-32 Screw Threads. Use with Single Cone Ferrules. Finger Tighten, 1 EA
Supplier:  Microsolv Technology Corporation 4732032
Encompass_Preferred
These Two Piece, "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. This fitting has a 12mm diameter knurled head, 18mm total length (top to bottom) and requires a Single Cone, Ferrule, sold separately. 2 Piece Male Nuts are often chosen because they can be used at slightly higher pressures compared to 1 Piece Nuts. The required, Single Coned Ferrules will swage tubing grip it and not spin as you turn the Nut. The larger Head makes it easier to produce the torque needed to achieve a good connection. As Ferrules wear down, they need to be replaced. This Nut can be reused with new Ferrules.
Catalog No. 50-227-1501