Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, One Piece, 1/16", Carbon PEEK, Black, Large Head with 10-32 Screw Threads. Finger Tighten, 10/PK.
Catalog No. 502271447
Encompass_Preferred
These "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. Fitting has a large diameter knurled head for ease of generating torque, 21mm total length (top to bottom) with a 12mm thread length. Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.
This item is not returnable. View return policy
Catalog No. 50-227-1447 Supplier Microsolv Technology Corporation Supplier No. 4730105
These "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. Fitting has a large diameter knurled head for ease of generating torque, 21mm total length (top to bottom) with a 12mm thread length. Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.