Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, One Piece, 1/16", PEEK, Natural, Small, "Combihead" Hex Head with 10-32 Screw Threads. Finger Tighten, 1 EA.
Catalog No. 502271496
Encompass_Preferred
This is one of our , "Male Nuts" used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. This unique Fitting has a 12mm diameter knurled head that has a hexagonal part for wrenching that final quarter turn or for lossening the nut. The smaller, 8mm diameter head allows this fitting to be used inside instruments when space is limited. The 28mm total length (top to bottom) with a 14mm thread length allows for a an excellent, swaged connection.
This item is not returnable. View return policy
Catalog No. 50-227-1496 Supplier Microsolv Technology Corporation Supplier No. 4732025
This is one of our , "Male Nuts" used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. This unique Fitting has a 12mm diameter knurled head that has a hexagonal part for wrenching that final quarter turn or for lossening the nut. The smaller, 8mm diameter head allows this fitting to be used inside instruments when space is limited. The 28mm total length (top to bottom) with a 14mm thread length allows for a an excellent, swaged connection.