Learn More
Microsolv Technology Corporation Tubing Connector Fittings, Low Pressure, 1/8", Carbon PEEK, Black with 1/4-28 Screw Threads, for use with Flange Free, polymeric tubing. 10/PK.
Supplier:  Microsolv Technology Corporation 4730301
Encompass_Preferred
These "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the low pressures zones of HPLC systems. This Two Piece Fitting must be used with a Ferrule. Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.
Catalog No. 50-227-1455