Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, One Piece, 1/16", PEEK, Natural, Large Head with 10-32 Screw Threads. Finger Tighten, 1 EA.
Catalog No. 502271481
Encompass_Preferred
This is one of our , "Male Nuts" used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. Fitting has a 12mm diameter knurled head, 21mm total length (top to bottom) with a 13mm thread length. The large head makes it easier to produce the torque needed to achieve a good swaged, connection.
This item is not returnable. View return policy
Catalog No. 50-227-1481 Supplier Microsolv Technology Corporation Supplier No. 4732005
This is one of our , "Male Nuts" used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. Fitting has a 12mm diameter knurled head, 21mm total length (top to bottom) with a 13mm thread length. The large head makes it easier to produce the torque needed to achieve a good swaged, connection.