Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, One Piece, 1/16", PEEK, Natural, Small Hex Head, Short Length and 10-32 Screw Threads. Wrench Tighten, 1 EA
Supplier:  Microsolv Technology Corporation 4732027
Encompass_Preferred
This "Male Nut" is used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. This Fitting has a 7mm diameter hexagonal head for use with a wrench to tighten or loosen the nut. The smaller, 7mm diameter head allows this fitting to be used inside instruments when space is limited. The 17mm total length (top to bottom) with a 13mm thread length allows for a an excellent, swaged connection that is similar in use to a Stainless Steel nut.
Catalog No. 50-227-1497