Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, Two Pieces, 1/16", PEEK, Natural, Small Hex Head, Standard Length and 10-32 Screw Threads. Use with Single Cone Ferrules. Wrench Tighten, 1 EA
Catalog No. 502271506
Encompass_Preferred
These Two Piece, "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. This fitting has a 7mm diameter knurled head, 15mm total length (top to bottom), 11mm Thread Length and requires a Single Cone Ferrule, sold separately. 2 Piece Male Nuts are often chosen because they can be used at slightly higher pressures compared to 1 Piece Nuts. The required, Single Coned Ferrules will swage tubing grip it and not spin as you turn the Nut. As Ferrules wear down, they need to be replaced. This Nut can be reused with new Ferrules.
This item is not returnable. View return policy
Catalog No. 50-227-1506 Supplier Microsolv Technology Corporation Supplier No. 4732040
These Two Piece, "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. This fitting has a 7mm diameter knurled head, 15mm total length (top to bottom), 11mm Thread Length and requires a Single Cone Ferrule, sold separately. 2 Piece Male Nuts are often chosen because they can be used at slightly higher pressures compared to 1 Piece Nuts. The required, Single Coned Ferrules will swage tubing grip it and not spin as you turn the Nut. As Ferrules wear down, they need to be replaced. This Nut can be reused with new Ferrules.