Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, Two Pieces, 1/16", PEEK, Natural, Small Hex Head, Short Length and 10-32 Screw Threads. Use with Double Cone Ferrules. Wrench Tighten, 10/PK.
Catalog No. 502271509
Encompass_Preferred
These Two Piece, "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. This fitting has a 7mm diameter knurled head, 11mm total length (top to bottom), 7mm Thread Length and requires a Double Cone Ferrule, sold separately. 2 Piece Male Nuts are often chosen because they can be used at slightly higher pressures compared to 1 Piece Nuts. The required, Single Coned Ferrules will swage tubing grip it and not spin as you turn the Nut. As Ferrules wear down, they need to be replaced. This Nut can be reused with new Ferrules.
This item is not returnable. View return policy
Catalog No. 50-227-1509 Supplier Microsolv Technology Corporation Supplier No. 4732043
These Two Piece, "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. This fitting has a 7mm diameter knurled head, 11mm total length (top to bottom), 7mm Thread Length and requires a Double Cone Ferrule, sold separately. 2 Piece Male Nuts are often chosen because they can be used at slightly higher pressures compared to 1 Piece Nuts. The required, Single Coned Ferrules will swage tubing grip it and not spin as you turn the Nut. As Ferrules wear down, they need to be replaced. This Nut can be reused with new Ferrules.