Learn More
Microsolv Technology Corporation Tubing Connector Fittings, Low Pressure, 1/8", Polypropylene, Yellow with 1/4-28 Screw Threads, for use with flanged, polymeric tubing. 10/PK.
Catalog No. 502271410
Encompass_Preferred
These "Male Nuts" are used to connect polymeric tubing in the low pressures zones of HPLC systems and other low pressure, fluidic uses. Tubing must be flanged and Washers are required to work properly, and sold separately.
This item is not returnable. View return policy
Catalog No. 50-227-1410 Supplier Microsolv Technology Corporation Supplier No. 45008Y
These "Male Nuts" are used to connect polymeric tubing in the low pressures zones of HPLC systems and other low pressure, fluidic uses. Tubing must be flanged and Washers are required to work properly, and sold separately.