Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, Two Pieces, 1/16", SS, Hex Heads with 10-32 Screw Threads and matching Single Cone Ferrules. Wrench Tighten, 10/PK.
Catalog No. 502270858
Encompass_Preferred
This is one of our , "Male Nuts" used to connect Stainless Steel tubing in the high pressures zones of HPLC systems. This economical 316L, Stainless Steel Fitting has a standard diameter Hexagonal head for wrenching the nut. The supplied 316L Stainless Steel matching Ferrule is designed to work with this fitting providing a permanent attachment to SS tubing for a high pressure connection.
This item is not returnable. View return policy
Catalog No. 50-227-0858 Supplier Microsolv Technology Corporation Supplier No. 4959010
This is one of our , "Male Nuts" used to connect Stainless Steel tubing in the high pressures zones of HPLC systems. This economical 316L, Stainless Steel Fitting has a standard diameter Hexagonal head for wrenching the nut. The supplied 316L Stainless Steel matching Ferrule is designed to work with this fitting providing a permanent attachment to SS tubing for a high pressure connection.