Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, White Polypropylene, Straight, Hose Barb Union, 4-5.25 mm ID; 100/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3060100
Encompass
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. NC9781671