Learn More
Ace Glass, Inc. Soft Copper Tubing for the ACE-Burlitch system.
SDP
Catalog No. 50979480
Encompass
soft copper tubing for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitably adapted conventional glassware.
Catalog No. 50-979-480 Supplier Ace Glass, Inc. Supplier No. 781858
soft copper tubing for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitably adapted conventional glassware.