Learn More
Trajan Scientific TUBING CUTTER 1/8IN
SDP
Catalog No. 502530189
Encompass_Preferred
SGE Tubing Cutter, For Use With: Used to get right angle cuts with minimal burring or chipping of the tubing, GC tubing, Stainless Steel
Catalog No. 50-253-0189 Supplier Trajan Scientific Supplier No. 082782
SGE Tubing Cutter, For Use With: Used to get right angle cuts with minimal burring or chipping of the tubing, GC tubing, Stainless Steel