Learn More
Microsolv Technology Corporation Tubing Cutter, Polymeric Tubing, Guillotine Style, Red. 1 EA.
Catalog No. 502271439
Encompass_Preferred
This low cost tubing cutter is intended for PTFE, ETFE, PFA, FEP and other soft polymeric tubing but can be used to cut PEEK tubing, but the cut tubing should not be used in critical HPLC connections.
This item is not returnable. View return policy
Catalog No. 50-227-1439 Supplier Microsolv Technology Corporation Supplier No. 4600201
This low cost tubing cutter is intended for PTFE, ETFE, PFA, FEP and other soft polymeric tubing but can be used to cut PEEK tubing, but the cut tubing should not be used in critical HPLC connections.