Learn More
Chemglass Life Sciences Tubing Cutter
SDP
Catalog No. CG197530
Encompass_Preferred
Tubing Cutter. For cutting tubing made from: Plastic, FEP, PTFE, Polyethylene, Polypropylene, or Rubber.
Catalog No. CG197530 Supplier Chemglass Life Sciences Supplier No. CG197530
Tubing Cutter. For cutting tubing made from: Plastic, FEP, PTFE, Polyethylene, Polypropylene, or Rubber.