Learn More
Accuflow Systems Saint-Gobain Tygon E-1000 Tubing, Non-DEHP, 1/2 in ID, 50 ft
Catalog No. NC3252491
Encompass_Preferred
Tygon E-1000 ultra-soft laboratory tubing is a non-DEHP formulation engineered for flexible fluid transfer in laboratory and analytical applications. The tubing remains flexible at very low temperatures, resists corrosive chemicals, and offers low compression resistance for peristaltic pump use.
  • Non-DEHP plasticizers.
  • 1/2 in (12.7 mm) internal diameter, 3/4 in (19.05 mm) outer diameter, 1/8 in (3.18 mm) wall thickness.
  • 50 ft packaged length.
  • Operating temperature: -67°F to 125°F (-55°C to 52°C).
  • Low durometer (≈40), suitable for low-torque peristaltic pumps.
  • Clear/semi-clear, compatible with many laboratory fluids and analytical instruments.
  • Maximum working pressure ~8 psi at 73°F; bend radius ~1.75 in.
Catalog No. NC3252491 Supplier Accuflow Systems Supplier No. ADK00038
Tygon E-1000 ultra-soft laboratory tubing is a non-DEHP formulation engineered for flexible fluid transfer in laboratory and analytical applications. The tubing remains flexible at very low temperatures, resists corrosive chemicals, and offers low compression resistance for peristaltic pump use.
  • Non-DEHP plasticizers.
  • 1/2 in (12.7 mm) internal diameter, 3/4 in (19.05 mm) outer diameter, 1/8 in (3.18 mm) wall thickness.
  • 50 ft packaged length.
  • Operating temperature: -67°F to 125°F (-55°C to 52°C).
  • Low durometer (≈40), suitable for low-torque peristaltic pumps.
  • Clear/semi-clear, compatible with many laboratory fluids and analytical instruments.
  • Maximum working pressure ~8 psi at 73°F; bend radius ~1.75 in.