Learn More
Microsolv Technology Corporation Tubing, ETFE, 0.020" ID, 1/16" OD, Low Pressure. 5 Meter Roll. 1 EA.
Catalog No. 502271520
Encompass_Preferred
This MicroSolv, ETFE ( Tefzel ) Tubing, is extruded from virgin ETFE and additives are not added to the original material. This tubing is used widely in all low-pressure, laboratory applications including before and after an HPLC Pump such as with Mobile Phase Delivery and Solvent Waste Systems. ETFE tubing is chemically inert except for some halogenated compounds and has low permeability compared with PTFE tubing. Internal and Outer Dimensional Tolerance: +/- 0.10mm Color: Natural ETFE, Translucent ID: 0.025" ( 0.51mm ) Pressure rating: 2175 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1520 Supplier Microsolv Technology Corporation Supplier No. 4851020
This MicroSolv, ETFE ( Tefzel ) Tubing, is extruded from virgin ETFE and additives are not added to the original material. This tubing is used widely in all low-pressure, laboratory applications including before and after an HPLC Pump such as with Mobile Phase Delivery and Solvent Waste Systems. ETFE tubing is chemically inert except for some halogenated compounds and has low permeability compared with PTFE tubing. Internal and Outer Dimensional Tolerance: +/- 0.10mm Color: Natural ETFE, Translucent ID: 0.025" ( 0.51mm ) Pressure rating: 2175 psi.