Learn More
Microsolv Technology Corporation Tubing Etfe 1/16 X.030 5M
Catalog No. 5001451612
Encompass_Preferred
Tubing, ETFE, 0.030 ID, 1/16 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51612 Supplier Microsolv Technology Corporation Supplier No. 4851030
Tubing, ETFE, 0.030 ID, 1/16 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.