Learn More
Microsolv Technology Corporation Tubing Etfe 1/16 X.040 5M
Catalog No. 5001451613
Encompass_Preferred
Tubing, ETFE, 0.040 ID, 1/16 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51613 Supplier Microsolv Technology Corporation Supplier No. 4851040
Tubing, ETFE, 0.040 ID, 1/16 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.