Learn More
Microsolv Technology Corporation Tubing, FEP, 0.010" ID, 1/16" OD, Low Pressure. 5 Meter Roll. 1 EA.
Catalog No. 502271518
Encompass_Preferred
This MicroSolv FEP ( Flourethylene Propylene ) low pressure tubing is chosen for use in Ion Chromatography Applications, but can be used in all low-pressure, laboratory applications including before and after an HPLC Pump such as with Mobile Phase Delivery and Solvent Waste Systems. Also known as Teflon FEP.Do not use this tubing with heat over 80⁰C. Internal and Outer Dimensional Tolerance: +/- 0.10mm Color: Natural FEP, Translucent ID: 0.010" (0.25 mm ) Pressure rating: 1650 psi.
This item is not returnable. View return policy
Catalog No. 50-227-1518 Supplier Microsolv Technology Corporation Supplier No. 4841010
This MicroSolv FEP ( Flourethylene Propylene ) low pressure tubing is chosen for use in Ion Chromatography Applications, but can be used in all low-pressure, laboratory applications including before and after an HPLC Pump such as with Mobile Phase Delivery and Solvent Waste Systems. Also known as Teflon FEP.Do not use this tubing with heat over 80⁰C. Internal and Outer Dimensional Tolerance: +/- 0.10mm Color: Natural FEP, Translucent ID: 0.010" (0.25 mm ) Pressure rating: 1650 psi.