Learn More
Microsolv Technology Corporation Tubing Fep 1/16 X.030 5M
Catalog No. 5001451608
Encompass_Preferred
Tubing, FEP, 0.030 ID, 1/16 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51608 Supplier Microsolv Technology Corporation Supplier No. 4841030
Tubing, FEP, 0.030 ID, 1/16 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.