Learn More
Microsolv Technology Corporation Tubing Fep 1/8 X.1/16Th 5M
Catalog No. 5001451610
Encompass_Preferred
Tubing, FEP, 1/16 ID, 1/8 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51610 Supplier Microsolv Technology Corporation Supplier No. 4844565
Tubing, FEP, 1/16 ID, 1/8 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.