Learn More
AVANTOR MASTERFLEX (VWR LLC) Flexelene SFX1.5-2.5N SFX Flexible Polyethylene Tubing, 3/32 x 5/32, 100' per pack
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9618002
Encompass
Applications: Pharmaceutical, biotech, and general lab applications. Characteristics: Good chemical resistance. Ultra-low extractables. Heat-sealable. Translucent. Certifications: Meets USP Class VI requirements. Temperature range: -40 to 100¿F (-40 to 37¿C) Sterilize: Ethylene oxide or gamma irradiation.
Catalog No. 50-630-279