Learn More
AVANTOR MASTERFLEX (VWR LLC) Flexelene SFX6-8N SFX Flexible Polyethylene Tubing, 3/8 x 1/2, 100' per pack
Catalog No. 50630285
Encompass
Applications: Pharmaceutical, biotech, and general lab applications. Characteristics: Good chemical resistance. Ultra-low extractables. Heat-sealable. Translucent. Certifications: Meets USP Class VI requirements. Temperature range: -40 to 100¿F (-40 to 37¿C) Sterilize: Ethylene oxide or gamma irradiation.
Catalog No. 50-630-285 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9618007
Applications: Pharmaceutical, biotech, and general lab applications. Characteristics: Good chemical resistance. Ultra-low extractables. Heat-sealable. Translucent. Certifications: Meets USP Class VI requirements. Temperature range: -40 to 100¿F (-40 to 37¿C) Sterilize: Ethylene oxide or gamma irradiation.