Learn More
Microsolv Technology Corporation Tubing Flx Peek .005 Id 50Mm R
Catalog No. 5001451659
Encompass_Preferred
Tubing, PEEK, Coiled, 0.005 ID, 1/16 OD. 1585mm L, General Grade, Red, with Tubing Connectors. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51659 Supplier Microsolv Technology Corporation Supplier No. 49308502R
Tubing, PEEK, Coiled, 0.005 ID, 1/16 OD. 1585mm L, General Grade, Red, with Tubing Connectors. MicroSolv Brand. 1/EA.