Learn More
Trajan Scientific Tubing Fused Silica FST Medical 1/EA
SDP
Supplier:  Trajan Scientific 064108
Encompass_Preferred
430 m OD x 320 m ID x 5 m length phenyl deactivated fused silica tubing
Catalog No. 50-004-93589