Learn More
Trajan Scientific Tubing Fused Silica FST Medical 1/EA
SDP
Catalog No. 5000494425
Encompass_Preferred
363 m OD x 220 m ID x 2 m length deactivated fused silica tubing
Catalog No. 50-004-94425 Supplier Trajan Scientific Supplier No. 0624469
363 m OD x 220 m ID x 2 m length deactivated fused silica tubing