Learn More
Trajan Scientific Tubing Fused Silica FST Medical 1/EA
SDP
Catalog No. 5000493989
Encompass_Preferred
363 m OD x 220 m ID x 5 m length deactivated fused silica tubing
Catalog No. 50-004-93989 Supplier Trajan Scientific Supplier No. 062447
363 m OD x 220 m ID x 5 m length deactivated fused silica tubing