Learn More
Trajan Scientific Tubing Fused Silica FST Medical 1/EA
SDP
Catalog No. 5000494040
Encompass_Preferred
680 m OD x 530 m ID x 25 m length deactivated fused silica tubing
Catalog No. 50-004-94040 Supplier Trajan Scientific Supplier No. 064034
680 m OD x 530 m ID x 25 m length deactivated fused silica tubing