Learn More
Trajan Scientific Tubing Fused Silica FST Medical 1/EA
SDP
Catalog No. 5000493745
Encompass_Preferred
680 m OD x 530 m ID x 5 m length deactivated fused silica tubing
Catalog No. 50-004-93745 Supplier Trajan Scientific Supplier No. 062448
680 m OD x 530 m ID x 5 m length deactivated fused silica tubing