Learn More
Trajan Scientific Tubing Fused Silica FST Medical 1/EA
SDP
Catalog No. 5000493945
Encompass_Preferred
150 m OD x 50 m ID x 10 m length fused silica tubing
Catalog No. 50-004-93945 Supplier Trajan Scientific Supplier No. 0624635
150 m OD x 50 m ID x 10 m length fused silica tubing