Learn More
Trajan Scientific Tubing Fused Silica FST Medical 1/EA
SDP
Catalog No. 5000494517
Encompass_Preferred
430 m OD x 320 m ID x 25 m length deactivated fused silica tubing
Catalog No. 50-004-94517 Supplier Trajan Scientific Supplier No. 0624473
430 m OD x 320 m ID x 25 m length deactivated fused silica tubing