Learn More
Trajan Scientific Tubing Fused Silica FST Medical 1/EA
SDP
Catalog No. 5000494496
Encompass_Preferred
285 m OD x 25 m ID x 10 m length fused silica tubing
Catalog No. 50-004-94496 Supplier Trajan Scientific Supplier No. 062460
285 m OD x 25 m ID x 10 m length fused silica tubing