Learn More
Trajan Scientific Tubing Fused Silica FST Medical 1/EA
SDP
Catalog No. 5000493658
Encompass_Preferred
430 m OD x 320 m ID x 5 m length deactivated fused silica tubing PK5
Catalog No. 50-004-93658 Supplier Trajan Scientific Supplier No. 064052
430 m OD x 320 m ID x 5 m length deactivated fused silica tubing PK5