Learn More
Trajan Scientific Tubing Fused Silica FST Medical 1/EA
SDP
Supplier:  Trajan Scientific 064102
Encompass_Preferred
363 m OD x 220 m ID x 5 m length phenyl deactivated fused silica tubing
Catalog No. 50-004-94917