Learn More
Trajan Scientific Tubing Fused Silica FST Medical 1/EA
SDP
Catalog No. 5000494415
Encompass_Preferred
363 m OD x 220 m ID x 10 m length fused silica tubing
Catalog No. 50-004-94415 Supplier Trajan Scientific Supplier No. 062475
363 m OD x 220 m ID x 10 m length fused silica tubing