Learn More
Trajan Scientific Tubing Fused Silica FST Medical 25/PK
SDP
Catalog No. 5000494931
Encompass_Preferred
363 m OD x 220 m ID x 5 m length deactivated fused silica tubing PK5
Catalog No. 50-004-94931 Supplier Trajan Scientific Supplier No. 064050
363 m OD x 220 m ID x 5 m length deactivated fused silica tubing PK5