Learn More
DSI LLC Tubing, Fused Silica untreated 360um x 150um, temperature resistant up to 350°C
Catalog No. 502255691
Encompass_Preferred
Tubing, Fused Silica untreated 360um x 150um, temperature resistant up to 350°C
Catalog No. 50-225-5691 Supplier DSI LLC Supplier No. JRT7360150
Tubing, Fused Silica untreated 360um x 150um, temperature resistant up to 350°C