Learn More
Trajan Scientific Tubing Glass lined GL Medical 1/EA
SDP
Catalog No. 5000493761
Encompass_Preferred
1/8 OD x 1 0 mm ID x 180 cm length Glass Lined Tubing
Catalog No. 50-004-93761 Supplier Trajan Scientific Supplier No. 082740
1/8 OD x 1 0 mm ID x 180 cm length Glass Lined Tubing