Learn More
Trajan Scientific Tubing Glass lined GL Medical 1/EA
SDP
Catalog No. 5000494257
Encompass_Preferred
1/8 OD x 0 5 mm ID x 30 cm length Glass Lined Tubing
Catalog No. 50-004-94257 Supplier Trajan Scientific Supplier No. 0827375
1/8 OD x 0 5 mm ID x 30 cm length Glass Lined Tubing