Learn More
Hach Company Tubing, Green/Green - PVC pump tubing replacement for Lachat FIA pumps (pack of 12)
Supplier:  Hach Company 53414
Encompass
PVC pump tubing supplied in a pack of 12 replacement tubes intended for flow injection analysis (FIA) and compatible pump applications. These green/green tubes are sold for use as replacement pump tubing in Lachat FIA systems and similar fluid-handling assemblies.
  • Material: PVC tubing supplied as replacement pump tubing.
  • Pack quantity: 12 tubes per package.
  • Intended application: Replacement pump tubing for flow injection analysis (FIA) systems.
  • Color: green/green tubes for easy identification.
Catalog No. NC2088485