Learn More
AVANTOR MASTERFLEX (VWR LLC) Tubing, High-Density Polyethylene (HDPE) , 3/8 x 1/2"", 100 ft/pack
Catalog No. NC1008252
Encompass
High-Density Polyethylene (HDPE) Tubing, 3/8 x 1/2", 100 ft/pack, Applications: General-purpose lab and industrial use.Characteristics: Low moisture absorption, good chemical and corrosion-resistance, high tensile strength.Certifications: None.Temperature range: ¿103 to 151¿F (¿75 to 66¿C)Sterilize: Autoclave.
Catalog No. NC1008252 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9550110
High-Density Polyethylene (HDPE) Tubing, 3/8 x 1/2", 100 ft/pack, Applications: General-purpose lab and industrial use.Characteristics: Low moisture absorption, good chemical and corrosion-resistance, high tensile strength.Certifications: None.Temperature range: ¿103 to 151¿F (¿75 to 66¿C)Sterilize: Autoclave.