Learn More
Waters Corp Waters Tubing Assembly, Head to Tee, Outlet, ISO 9001, HPLC
Catalog No. 50818809 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly functions as an outlet connection from the pump head to the tee in various High-Performance Liquid Chromatography (HPLC) systems. It is designed as a high-quality, direct replacement component, ensuring seamless integration without modifications. The assembly is manufactured to stringent ISO 9001 quality standards.
  • Functions as outlet tubing for left and right connections
  • Compatible with a range of HPLC systems
  • Manufactured to ISO 9001 quality standards
  • Direct drop-in replacement, requiring no modifications
  • Supplied as one unit per package
Catalog No. 50-818-809 Supplier Waters Corp Supplier No. WAT060069
This tubing assembly functions as an outlet connection from the pump head to the tee in various High-Performance Liquid Chromatography (HPLC) systems. It is designed as a high-quality, direct replacement component, ensuring seamless integration without modifications. The assembly is manufactured to stringent ISO 9001 quality standards.
  • Functions as outlet tubing for left and right connections
  • Compatible with a range of HPLC systems
  • Manufactured to ISO 9001 quality standards
  • Direct drop-in replacement, requiring no modifications
  • Supplied as one unit per package